Learn More
Medchemexpress LLC HY-105008 5mg Medchemexpress, Secoisolariciresinol diglucoside CAS:257930-74-8 Purity:>98%
SDP
Catalog No. 501968240
Encompass_Preferred
Medchemexpress, HY-105008 5mg Secoisolariciresinol diglucoside CAS:257930-74-8 Secoisolariciresinol diglucoside ((S,S)-SDG) is a major component of the lignans in flaxseed, scavenges free radical, with potent antioxidant activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8240 Supplier Medchemexpress LLC Supplier No. HY1050085MG
Medchemexpress, HY-105008 5mg Secoisolariciresinol diglucoside CAS:257930-74-8 Secoisolariciresinol diglucoside ((S,S)-SDG) is a major component of the lignans in flaxseed, scavenges free radical, with potent antioxidant activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.