Learn More
Cayman Chemical Secoisolariciresinol Diglucoside, Purity: ≥98%, CAS: 158932-33-3, 100 mg
SDP
Catalog No. 502933621
Encompass_Preferred
A lignan with diverse biological activities; scavenges DPPH radicals (IC50 = 78.9 μg/ml) in a cell-free assay; dose-dependently inhibits the growth of SW480 human colon cancer cells; dietary administration reduces the number of proliferating tumor cells in an MCF-7 human breast cancer mouse xenograft model in the presence of circulating estrogen; increases insulin and decreases glucose serum levels in rats with streptozotocin-induced diabetes at 20 mg/kg; decreases infarct size in rats with myocardial infarction induced by permanent occlusion of the left anterior descending coronary artery at a 20 mg/kg per day
Catalog No. 50-293-3621 Supplier Cayman Chemical Supplier No. 24974100 MG
A lignan with diverse biological activities; scavenges DPPH radicals (IC50 = 78.9 μg/ml) in a cell-free assay; dose-dependently inhibits the growth of SW480 human colon cancer cells; dietary administration reduces the number of proliferating tumor cells in an MCF-7 human breast cancer mouse xenograft model in the presence of circulating estrogen; increases insulin and decreases glucose serum levels in rats with streptozotocin-induced diabetes at 20 mg/kg; decreases infarct size in rats with myocardial infarction induced by permanent occlusion of the left anterior descending coronary artery at a 20 mg/kg per day