Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Secologanoside | 59472-23-0 | MFCD32201171 | 5 MG
SDP

Catalog No. 5000417771
Encompass_Preferred

Secologanoside is a triterpenoid isolated from Poraqueiba sericea. It weakly inhibits elastase with an IC50 of 164 μg/mL and is moderately cytotoxic to fibroblasts. It is for research use only.

  • Weakly inhibits elastase
  • Moderately cytotoxic to fibroblasts
  • High purity (99.0%)
  • Molecular formula C16H22O11
  • Appearance: white to off-white solid
  • Soluble in H2O (25 mg/mL)

Catalog No. 50-004-17771 Supplier Medchemexpress LLC Supplier No. HYN68765MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Secologanoside is a triterpenoid isolated from Poraqueiba sericea. It weakly inhibits elastase with an IC50 of 164 μg/mL and is moderately cytotoxic to fibroblasts. It is for research use only.

  • Weakly inhibits elastase
  • Moderately cytotoxic to fibroblasts
  • High purity (99.0%)
  • Molecular formula C16H22O11
  • Appearance: white to off-white solid
  • Soluble in H2O (25 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.