Learn More
Cytek Biosciences Inc Second Step Reagents APC Streptavidin ..... Cat. No. 20-4317-U500 ..... Size 500 µg
Catalog No. 502102763
Encompass_Preferred
Second Step Reagents APC Streptavidin ..... Cat. No. 20-4317-U500 ..... Size 500 µgAPC Streptavidin is a second-step reagent useful when detecting biotinylated antibodies in indirect staining protocols. Streptavidin is a nonglycosylated biotin-binding protein that is bacterially derived. Like avidin, it binds to biotin with high affinity but displays less nonspecific binding.
Catalog No. 50-210-2763 Supplier Cytek Biosciences Inc Supplier No. 204317U500
Second Step Reagents APC Streptavidin ..... Cat. No. 20-4317-U500 ..... Size 500 µgAPC Streptavidin is a second-step reagent useful when detecting biotinylated antibodies in indirect staining protocols. Streptavidin is a nonglycosylated biotin-binding protein that is bacterially derived. Like avidin, it binds to biotin with high affinity but displays less nonspecific binding.