Learn More
Thomas Scientific Secondary Containment Tray, 34 x 9 x 1"
SDP
Catalog No. NC1488781
Encompass
Secondary contaiment is required by OSHA and EPA in chemical storerrooms and should be a part of every lab Chemical Hygiene Program. Laboratory spill trays are a minimum 3/16” thick, one piece stress free, virgin polyethylene with smooth interior walls and coved corners for easy cleaning. Trays conform to ASTM #1412 for corrosion resistance, and provide secondary containment complying with OSHA 1910.1450 and EPA 264.175 for chemical stockrooms/storerooms All trays are manufactured in the USA
Catalog No. NC1488781 Supplier Thomas Scientific Supplier No. 1185F69
Secondary contaiment is required by OSHA and EPA in chemical storerrooms and should be a part of every lab Chemical Hygiene Program. Laboratory spill trays are a minimum 3/16” thick, one piece stress free, virgin polyethylene with smooth interior walls and coved corners for easy cleaning. Trays conform to ASTM #1412 for corrosion resistance, and provide secondary containment complying with OSHA 1910.1450 and EPA 264.175 for chemical stockrooms/storerooms All trays are manufactured in the USA