Learn More
Thomas Scientific Secondary Containment Tray, 17 x 9 x 1"
SDP
Supplier:  Thomas Scientific 1185F68
Encompass
Secondary contaiment is required by OSHA and EPA in chemical storerrooms and should be a part of every lab Chemical Hygiene Program. Laboratory spill trays are a minimum 3/16” thick, one piece stress free, virgin polyethylene with smooth interior walls and coved corners for easy cleaning. Trays conform to ASTM #1412 for corrosion resistance, and provide secondary containment complying with OSHA 1910.1450 and EPA 264.175 for chemical stockrooms/storerooms All trays are manufactured in the USA
Catalog No. NC1488783