Learn More
Cell Signaling Technology Anti-Biotin (D5A7) Rabbit Monoclonal Antibody, HRP Conjugate, All Species Reactivity, 1 mL
SDP
Catalog No. 50195910
Encompass
The Anti-biotin (D5A7) Rabbit monoclonal antibody is conjugated to the carbohydrate groups of horseradish peroxidase (HRP) via its amine groups. This conjugate is optimized for the detection of biotinylated primary antibodies, eliminating the need for incubation with a secondary antibody in many applications.
  • Recognizes biotin attached to proteins, peptides, oligonucleotides and solid matrices.
  • Optimized for peptide ELISA and western blotting.
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 2 mg/mL bovine serum albumin, and 50% glycerol.
  • Validated for all species reactivity.
Catalog No. 50-195-910 Supplier Cell Signaling Technology Supplier No. 5571S
The Anti-biotin (D5A7) Rabbit monoclonal antibody is conjugated to the carbohydrate groups of horseradish peroxidase (HRP) via its amine groups. This conjugate is optimized for the detection of biotinylated primary antibodies, eliminating the need for incubation with a secondary antibody in many applications.
  • Recognizes biotin attached to proteins, peptides, oligonucleotides and solid matrices.
  • Optimized for peptide ELISA and western blotting.
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 2 mg/mL bovine serum albumin, and 50% glycerol.
  • Validated for all species reactivity.