Learn More
Cell Signaling Technology Anti-Mouse IgG, HRP-Linked Antibody, Horse Host, 1 mL
SDP
Catalog No. 50195914
Encompass
This affinity-purified horse anti-mouse IgG (heavy and light chain) antibody is conjugated to horseradish peroxidase (HRP) for efficient chemiluminescent detection. This reagent is validated for use with primary antibodies to ensure reproducible results in western immunoblotting applications.
  • Conjugated to HRP for chemiluminescent detection
  • Validated for western blotting applications
  • Suitable for detecting proteins in mouse-derived samples
  • Supplied in 10mM sodium HEPES (pH 7.5), 150 mM NaCl, 2 mg/mL bovine serum albumin (BSA), and 50% glycerol
  • Stable for 60 months when stored at -20°C
Catalog No. 50-195-914 Supplier Cell Signaling Technology Supplier No. 7076S
This affinity-purified horse anti-mouse IgG (heavy and light chain) antibody is conjugated to horseradish peroxidase (HRP) for efficient chemiluminescent detection. This reagent is validated for use with primary antibodies to ensure reproducible results in western immunoblotting applications.
  • Conjugated to HRP for chemiluminescent detection
  • Validated for western blotting applications
  • Suitable for detecting proteins in mouse-derived samples
  • Supplied in 10mM sodium HEPES (pH 7.5), 150 mM NaCl, 2 mg/mL bovine serum albumin (BSA), and 50% glycerol
  • Stable for 60 months when stored at -20°C