Learn More
Zymo Research Corporation Secondary Antibody 30 ul
SDP
Catalog No. 502037154
Encompass_Preferred
Secondary Antibody 15ul/30ul @1ug/ul. For use in the 5-mC DNA ELISA kit for DNA methylation quantification. Can be used for other applications requring 5-mC antibodies.
Catalog No. 50-203-7154 Supplier Zymo Research Corporation Supplier No. D5325330
Secondary Antibody 15ul/30ul @1ug/ul. For use in the 5-mC DNA ELISA kit for DNA methylation quantification. Can be used for other applications requring 5-mC antibodies.