Learn More
Biotium Secondary Antibody Alpaca IgG VHH domain Llama IgG VHH domain Goat CF488A 1/EA
Catalog No. 5000500661
Encompass_Preferred
High-quality goat anti-alpaca IgG VHH labeled with green fluorescent CF488A dye (Ex/Em 490/515 nm) CF dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes
Catalog No. 50-005-00661 Supplier Biotium Supplier No. 20882100UL
High-quality goat anti-alpaca IgG VHH labeled with green fluorescent CF488A dye (Ex/Em 490/515 nm) CF dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes