Learn More
Biotium Secondary Antibody Alpaca IgG VHH domain Llama IgG VHH domain Goat CF568 1/EA
Catalog No. 5000501267
Encompass_Preferred
High-quality goat anti-alpaca IgG VHH labeled with red fluorescent CF568 dye (Ex/Em 562/583 nm) CF dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes
Catalog No. 50-005-01267 Supplier Biotium Supplier No. 20883100UL
High-quality goat anti-alpaca IgG VHH labeled with red fluorescent CF568 dye (Ex/Em 562/583 nm) CF dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes