Learn More
Biotium Secondary Antibody Alpaca IgG VHH domain Llama IgG VHH domain Goat CF594 1/EA
Catalog No. 5000499414
Encompass_Preferred
High-quality goat anti-alpaca IgG VHH labeled with deep-red fluorescent CF594 dye (Ex/Em 593/615 nm) CF dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes
Catalog No. 50-004-99414 Supplier Biotium Supplier No. 20884100UL
High-quality goat anti-alpaca IgG VHH labeled with deep-red fluorescent CF594 dye (Ex/Em 593/615 nm) CF dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes