Learn More
Biotium Secondary Antibody Alpaca IgG VHH domain Llama IgG VHH domain Goat CF647 1/EA
Catalog No. 5000499904
Encompass_Preferred
High-quality goat anti-alpaca IgG VHH labeled with far-red fluorescent CF647 dye (Ex/Em 650/665 nm) CF dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes
Catalog No. 50-004-99904 Supplier Biotium Supplier No. 20886100UL
High-quality goat anti-alpaca IgG VHH labeled with far-red fluorescent CF647 dye (Ex/Em 650/665 nm) CF dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes