Learn More
Biotium Secondary Antibody Alpaca IgG VHH domain Llama IgG VHH domain Goat CF680 1/EA
Catalog No. 5000500123
Encompass_Preferred
High-quality goat anti-alpaca IgG VHH labeled with far-red/near-IR CF680R dye (Ex/Em 680/701 nm) CF dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes
Catalog No. 50-005-00123 Supplier Biotium Supplier No. 20887100UL
High-quality goat anti-alpaca IgG VHH labeled with far-red/near-IR CF680R dye (Ex/Em 680/701 nm) CF dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes