Learn More
Biotium Secondary Antibody FITC 1F8.1E4 CF405M 1/EA
Catalog No. 5000499410
Encompass_Preferred
This product is prepared by labeling monoclonal mouse anti-fluorescein with CF and reg 405M dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Blue fluorescent CF and reg 405M dye has excitation/emission at 408/452 nm Note Conjugates of blue fluorescent dyes like CF and reg 350 CF and reg 405S and CF and reg 405M are not recommended for detecting low abundance targets because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors
Catalog No. 50-004-99410 Supplier Biotium Supplier No. 20214
Add to Cart
Add to Cart
This product is prepared by labeling monoclonal mouse anti-fluorescein with CF and reg 405M dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Blue fluorescent CF and reg 405M dye has excitation/emission at 408/452 nm Note Conjugates of blue fluorescent dyes like CF and reg 350 CF and reg 405S and CF and reg 405M are not recommended for detecting low abundance targets because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors