Learn More
Biotium Secondary Antibody FITC 1F8.1E4 CF594 1/EA
Catalog No. 5000499444
Encompass_Preferred
This product is prepared by labeling monoclonal mouse anti-fluorescein with CF and reg 594 dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Deep-red fluorescent CF and reg 594 dye has excitation/emission at 593/614 nm
Catalog No. 50-004-99444 Supplier Biotium Supplier No. 20211
Add to Cart
Edge
Add to Cart
This product is prepared by labeling monoclonal mouse anti-fluorescein with CF and reg 594 dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Deep-red fluorescent CF and reg 594 dye has excitation/emission at 593/614 nm