Learn More
Biotium Secondary Antibody GFP 9F9.F9 CF633 1/EA
Catalog No. 5000501265
Encompass_Preferred
This product is prepared by labeling monoclonal mouse anti-green fluorescent protein (GFP) with CF and reg 633 dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Far-red fluorescent CF and reg 633 dye has excitation/emission at 630/650 nm
Catalog No. 50-005-01265 Supplier Biotium Supplier No. 20217
Add to Cart
Edge
Add to Cart
This product is prepared by labeling monoclonal mouse anti-green fluorescent protein (GFP) with CF and reg 633 dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Far-red fluorescent CF and reg 633 dye has excitation/emission at 630/650 nm