Learn More
Biotium Secondary Antibody Goat IgG Bovine CF543 1/EA
Catalog No. 5000500595
Encompass_Preferred
This product is bovine anti-goat IgG labeled with CF and reg 543 dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Orange fluorescent CF and reg 543 dye has excitation/emission at 541/560 nm The antibody is created by immunizing a cow host with goat IgG to minimize the reactivity for bovine IgG The antibody has been adsorbed against bovine chicken guinea pig Syrian hamster horse human mouse rabbit and rat serum proteins
Catalog No. 50-005-00595 Supplier Biotium Supplier No. 20313
This product is bovine anti-goat IgG labeled with CF and reg 543 dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Orange fluorescent CF and reg 543 dye has excitation/emission at 541/560 nm The antibody is created by immunizing a cow host with goat IgG to minimize the reactivity for bovine IgG The antibody has been adsorbed against bovine chicken guinea pig Syrian hamster horse human mouse rabbit and rat serum proteins