Learn More
Biotium Secondary Antibody Goat IgG Bovine CF633 1/EA
Catalog No. 5000501173
Encompass_Preferred
This product is bovine anti-goat IgG labeled with CF and reg 594ST dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Deep-red fluorescent CF and reg 594ST dye has excitation/emission at 593/614 nm and is designed specifically for STORM super-resolution imaging The antibody is created by immunizing a cow host with goat IgG to minimize the reactivity for bovine IgG The antibody has been adsorbed against bovine chicken guinea pig Syrian hamster horse human mouse rabbit and rat serum proteins
Catalog No. 50-005-01173 Supplier Biotium Supplier No. 20296
This product is bovine anti-goat IgG labeled with CF and reg 594ST dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Deep-red fluorescent CF and reg 594ST dye has excitation/emission at 593/614 nm and is designed specifically for STORM super-resolution imaging The antibody is created by immunizing a cow host with goat IgG to minimize the reactivity for bovine IgG The antibody has been adsorbed against bovine chicken guinea pig Syrian hamster horse human mouse rabbit and rat serum proteins