Learn More
Biotium Secondary Antibody Goat IgG Donkey CF505 1/EA
Catalog No. 5000499902
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed donkey anti-goat IgG (H plus L) with CF and reg 505 dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Red fluorescent CF and reg 505 dye has excitation/emission at 505/519 nm To minimize cross-reactivity the antibody has been adsorbed against chicken guinea pig Syrian hamster horse human mouse rabbit and rat serum Single label antibodies have an average of one dye molecule per antibody molecule which is reported to be optimal for super-resolution imaging by STORM
Catalog No. 50-004-99902 Supplier Biotium Supplier No. 2088050UL
This product is prepared by labeling highly cross-adsorbed donkey anti-goat IgG (H plus L) with CF and reg 505 dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Red fluorescent CF and reg 505 dye has excitation/emission at 505/519 nm To minimize cross-reactivity the antibody has been adsorbed against chicken guinea pig Syrian hamster horse human mouse rabbit and rat serum Single label antibodies have an average of one dye molecule per antibody molecule which is reported to be optimal for super-resolution imaging by STORM