Learn More
Biotium Secondary Antibody Goat IgG Donkey CF740 1/EA
Supplier:  Biotium 2098750UL
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed donkey anti-goat IgG (H plus L) with CF and reg 740 dye To minimize cross-reactivity the antibody has been adsorbed against chicken guinea pig Syrian hamster horse human mouse rabbit and rat serum
Catalog No. 50-005-01329