Learn More
Biotium Secondary Antibody Guinea Pig IgG Donkey CF505 1/EA
Catalog No. 5000500097
Encompass_Preferred
Highly cross-adsorbed donkey anti-guinea pig IgG (H plus L) labeled with CF and reg 505 dye Single CF dye label (DOL equals 1) optimized for super-resolution imaging by STORM Adsorbed against bovine chicken goat Syrian hamster horse human mouse rabbit sheep and rat serum
Catalog No. 50-005-00097 Supplier Biotium Supplier No. 2088150UL
Highly cross-adsorbed donkey anti-guinea pig IgG (H plus L) labeled with CF and reg 505 dye Single CF dye label (DOL equals 1) optimized for super-resolution imaging by STORM Adsorbed against bovine chicken goat Syrian hamster horse human mouse rabbit sheep and rat serum