Learn More
Biotium Secondary Antibody Guinea Pig IgG Donkey CF583R 1/EA
Catalog No. 5000500210
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed donkey anti-guinea pig IgG (H plus L) with CF and reg 583R dye To minimize cross-reactivity the antibody has been adsorbed against bovine chicken goat Syrian hamster horse human mouse rabbit sheep and rat serum
Catalog No. 50-005-00210 Supplier Biotium Supplier No. 2089350UL
This product is prepared by labeling highly cross-adsorbed donkey anti-guinea pig IgG (H plus L) with CF and reg 583R dye To minimize cross-reactivity the antibody has been adsorbed against bovine chicken goat Syrian hamster horse human mouse rabbit sheep and rat serum