Learn More
Biotium Secondary Antibody Mouse IgG Donkey CF583R 1/EA
Catalog No. 5000500120
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed donkey anti-mouse IgG (H plus L) with CF and reg 583R dye To minimize cross-reactivity the antibody has been adsorbed against bovine chicken goat guinea pig Syrian hamster horse human rabbit and sheep serum
Catalog No. 50-005-00120 Supplier Biotium Supplier No. 20894500UL
This product is prepared by labeling highly cross-adsorbed donkey anti-mouse IgG (H plus L) with CF and reg 583R dye To minimize cross-reactivity the antibody has been adsorbed against bovine chicken goat guinea pig Syrian hamster horse human rabbit and sheep serum