Learn More
Biotium Secondary Antibody Mouse IgG Goat Biotin 1/EA
Catalog No. 5000501178
Encompass_Preferred
This product is prepared by labeling goat anti-mouse IgG3 with biotin To minimize cross reactivity the antibodies are cross-adsorbed against other mouse IgG subclasses (IgG1 IgG2a IgG2b IgG2c) and human bovine and rabbit serum proteins
Catalog No. 50-005-01178 Supplier Biotium Supplier No. 2092150UL
This product is prepared by labeling goat anti-mouse IgG3 with biotin To minimize cross reactivity the antibodies are cross-adsorbed against other mouse IgG subclasses (IgG1 IgG2a IgG2b IgG2c) and human bovine and rabbit serum proteins