Learn More
Biotium Secondary Antibody Mouse IgG Goat CF488A 1/EA
Catalog No. 5000501307
Encompass_Preferred
Goat anti-mouse IgG3 labeled with CF and reg 488A dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes To minimize cross reactivity the antibodies are cross-adsorbed against other mouse IgG subclasses (IgG1 IgG2a IgG2b IgG2c) and human bovine and rabbit serum proteins
Catalog No. 50-005-01307 Supplier Biotium Supplier No. 2092450UL
Goat anti-mouse IgG3 labeled with CF and reg 488A dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes To minimize cross reactivity the antibodies are cross-adsorbed against other mouse IgG subclasses (IgG1 IgG2a IgG2b IgG2c) and human bovine and rabbit serum proteins