Learn More
Biotium Secondary Antibody Mouse IgG Llama CF583R 1/EA
Catalog No. 5000501071
Encompass_Preferred
This product is prepared by labeling an affinity-purified llama anti-mouse IgG with CF and reg 583R dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes
Catalog No. 50-005-01071 Supplier Biotium Supplier No. 2090850UL
This product is prepared by labeling an affinity-purified llama anti-mouse IgG with CF and reg 583R dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes