Learn More
Biotium Secondary Antibody Mouse IgG Llama CF583R 1/EA
Supplier: Biotium 20908500UL
This product is prepared by labeling an affinity-purified llama anti-mouse IgG with CF and reg 583R dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes