Learn More
Biotium Secondary Antibody Mouse IgG Rabbit CF583R 1/EA
Supplier:  Biotium 20911500UL
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed rabbit anti-mouse IgG (H plus L) with CF and reg 583R dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes
Catalog No. 50-005-00173