Learn More
Biotium Secondary Antibody Mouse IgG Rabbit CF583R 1/EA
Catalog No. 5000501227
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed rabbit anti-mouse IgG (H plus L) with CF and reg 583R dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes
Catalog No. 50-005-01227 Supplier Biotium Supplier No. 2091150UL
This product is prepared by labeling highly cross-adsorbed rabbit anti-mouse IgG (H plus L) with CF and reg 583R dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes