Learn More
Biotium Secondary Antibody Rabbit IgG Chicken CF583R 1/EA
Catalog No. 5000500212
Encompass_Preferred
This product is prepared by labeling an affinity-purified chicken anti-rabbit IgG (H plus L) with CF and reg 583R dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes
Catalog No. 50-005-00212 Supplier Biotium Supplier No. 20891500UL
This product is prepared by labeling an affinity-purified chicken anti-rabbit IgG (H plus L) with CF and reg 583R dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes