Learn More
Biotium Secondary Antibody Rabbit IgG Donkey CF505 1/EA
Supplier: Biotium 20879500UL
This product is prepared by labeling highly cross-adsorbed donkey anti-rabbit IgG (H plus L) with CF and reg 505 dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Red fluorescent CF and reg 505 dye has excitation/emission at 505/519 nm To minimize cross-reactivity the antibody has been adsorbed against bovine chicken goat guinea pig Syrian hamster horse human mouse rat and sheep serum Single label antibodies have an average of one dye molecule per antibody molecule which is reported to be optimal for super-resolution imaging by STORM