Learn More
Biotium Secondary Antibody Rabbit IgG Donkey CF660R 1/EA
Catalog No. 5000501586
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed donkey anti-rabbit IgG (H plus L) with CF and reg 660R dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Far-red fluorescent CF and reg 660R dye has excitation/emission at 663/682 nm To minimize cross-reactivity the antibody has been adsorbed against bovine chicken goat guinea pig Syrian hamster horse human mouse rat and sheep serum
Catalog No. 50-005-01586 Supplier Biotium Supplier No. 2038950UL
This product is prepared by labeling highly cross-adsorbed donkey anti-rabbit IgG (H plus L) with CF and reg 660R dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Far-red fluorescent CF and reg 660R dye has excitation/emission at 663/682 nm To minimize cross-reactivity the antibody has been adsorbed against bovine chicken goat guinea pig Syrian hamster horse human mouse rat and sheep serum