Learn More
Biotium Secondary Antibody Rabbit IgG Donkey CF740 1/EA
Catalog No. 5000500161
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed donkey anti-rabbit IgG (H plus L) with CF and reg 740 dye To minimize cross-reactivity the antibody has been adsorbed against bovine chicken goat guinea pig Syrian hamster horse human mouse rat and sheep serum
Catalog No. 50-005-00161 Supplier Biotium Supplier No. 2098650UL
This product is prepared by labeling highly cross-adsorbed donkey anti-rabbit IgG (H plus L) with CF and reg 740 dye To minimize cross-reactivity the antibody has been adsorbed against bovine chicken goat guinea pig Syrian hamster horse human mouse rat and sheep serum