Learn More
Biotium Secondary Antibody Rabbit IgG Goat CF740 1/EA
Supplier:  Biotium 20990250UL
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed goat anti-rabbit IgG (H plus L) with CF and reg 740 dye CF and reg 740 dye has excitation/emission at 740/775 nm To minimize cross-reactivity the antibody has been adsorbed against human mouse and rat serum
Catalog No. 50-005-00254