Learn More
Biotium Secondary Antibody Rabbit IgG Goat CF740 1/EA
Catalog No. 5000501076
Encompass_Preferred
This product is prepared by labeling high quality goat anti-rabbit IgG (H plus L) with CF and reg 740 dye CF and reg 740 dye has excitation/emission at 740/775 nm
Catalog No. 50-005-01076 Supplier Biotium Supplier No. 2098550UL
This product is prepared by labeling high quality goat anti-rabbit IgG (H plus L) with CF and reg 740 dye CF and reg 740 dye has excitation/emission at 740/775 nm