Learn More
Biotium Secondary Antibody Rabbit IgG Goat CF740 1/EA
Supplier:  Biotium 20985500UL
Encompass_Preferred
This product is prepared by labeling high quality goat anti-rabbit IgG (H plus L) with CF and reg 740 dye CF and reg 740 dye has excitation/emission at 740/775 nm
Catalog No. 50-005-01237