Learn More
Biotium Secondary Antibody Rabbit IgG Goat CF750 1/EA
Catalog No. 5000501179
Encompass_Preferred
This product is prepared by labeling high quality F(ab)2 fragment of goat anti-rabbit IgG (H plus L) with CF and reg 750 dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes
Catalog No. 50-005-01179 Supplier Biotium Supplier No. 20919250UL
This product is prepared by labeling high quality F(ab)2 fragment of goat anti-rabbit IgG (H plus L) with CF and reg 750 dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes