Learn More
Biotium Secondary Antibody Rat IgG Donkey CF740 1/EA
Supplier:  Biotium 2098950UL
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed donkey anti-rat IgG (H plus L) with CF and reg 740 dye To minimize cross-reactivity the antibody has been adsorbed against bovine chicken goat guinea pig Syrian hamster horse human mouse rabbit and sheep serum proteins
Catalog No. 50-005-00162