Learn More
Biotium Secondary Antibody Rat IgG Rabbit CF583R 1/EA
Supplier:  Biotium 20912500UL
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed rabbit anti-rat IgG (H plus L) with CF and reg 583R dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes
Catalog No. 50-005-01304