Learn More
Biotium Secondary Antibody Sheep IgG Donkey CF583R 1/EA
Catalog No. 5000500126
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed donkey anti-sheep IgG (H plus L) with CF and reg 583R dye To minimize cross-reactivity the antibody has been adsorbed against chicken guinea pig Syrian hamster horse human mouse rabbit and rat serum
Catalog No. 50-005-00126 Supplier Biotium Supplier No. 2089750UL
This product is prepared by labeling highly cross-adsorbed donkey anti-sheep IgG (H plus L) with CF and reg 583R dye To minimize cross-reactivity the antibody has been adsorbed against chicken guinea pig Syrian hamster horse human mouse rabbit and rat serum