Learn More
Foxx Life Sciences EZwaste Secondary Container Spill Basin, Safety Tray for 10L-20L (2.5 to 5 Gallon) Carboys, Bottles, and DOT Waste Containers, 6/PK
Catalog No. 501178530
Encompass_Preferred
252-1906-OEM: EZwaster Safety Tray Secondary Container, for 10L-20L Carboys, 6/pk . Foxx Life Sciences' Secondary Container is a perfect solution for preventing leakage and spillage from Foxx Life Sciences 13.5L and 20L EZwaste Solvent Waste System containers. Made from HDPE material, these containers are built to be tough and durable. Our Secondary Containers have the ability to hold at least 115% of the container content.
Catalog No. 50-117-8530 Supplier Foxx Life Sciences Supplier No. 2521906OEM
252-1906-OEM: EZwaster Safety Tray Secondary Container, for 10L-20L Carboys, 6/pk . Foxx Life Sciences' Secondary Container is a perfect solution for preventing leakage and spillage from Foxx Life Sciences 13.5L and 20L EZwaste Solvent Waste System containers. Made from HDPE material, these containers are built to be tough and durable. Our Secondary Containers have the ability to hold at least 115% of the container content.