Learn More
Foxx Life Sciences EZwaste Secondary Container Spill Basin, Safety Tray for 20L-60L (5 to 15 Gallon) Carboys, Bottles, and DOT Waste Containers
Catalog No. 501178535
Encompass_Preferred
252-1903-OEM: EZwaste Safety Tray Secondary Container, for 20L-60L Carboys . Foxx Life Sciences' Secondary Container is a perfect solution for preventing leakage and spillage from Foxx Life Sciences 13.5L and 20L EZwaste Solvent Waste System containers. Made from HDPE material, these containers are built to be tough and durable. Our Secondary Containers have the ability to hold at least 115% of the container content.
Catalog No. 50-117-8535 Supplier Foxx Life Sciences Supplier No. 2521903OEM
252-1903-OEM: EZwaste Safety Tray Secondary Container, for 20L-60L Carboys . Foxx Life Sciences' Secondary Container is a perfect solution for preventing leakage and spillage from Foxx Life Sciences 13.5L and 20L EZwaste Solvent Waste System containers. Made from HDPE material, these containers are built to be tough and durable. Our Secondary Containers have the ability to hold at least 115% of the container content.