Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Secoxyloganin | 58822-47-2 | MFCD20260822 | 99.3% | 404.37 g/mol | C17H24O11 | 10 MG
SDP

Catalog No. 5000289417
Encompass_Preferred

Secoxyloganin is an iridoid glycoside isolated from Lonicera japonica and provided for research and analytical use. It is studied for allergy-preventive activity and its effects on blood flow. The compound is a white to light-yellow solid with a molecular weight of 404.37 g/mol and high purity suitable for analytical applications.

  • High purity: 99.3% suitable for analytical use.
  • Available in multiple pack sizes, including 10 MG.
  • Solvent stability: store in solvent at -80°C for long-term, -20°C for short-term.
  • Appearance: white to light yellow solid.
  • Intended for research and analytical applications only (not for human use).

Catalog No. 50-002-89417 Supplier Medchemexpress LLC Supplier No. HYN300910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Secoxyloganin is an iridoid glycoside isolated from Lonicera japonica and provided for research and analytical use. It is studied for allergy-preventive activity and its effects on blood flow. The compound is a white to light-yellow solid with a molecular weight of 404.37 g/mol and high purity suitable for analytical applications.

  • High purity: 99.3% suitable for analytical use.
  • Available in multiple pack sizes, including 10 MG.
  • Solvent stability: store in solvent at -80°C for long-term, -20°C for short-term.
  • Appearance: white to light yellow solid.
  • Intended for research and analytical applications only (not for human use).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.