Learn More
Cayman Chemical Secoxyloganin, Purity: ≥95%, CAS: 58822-47-2, 10 mg
SDP
Catalog No. 502988958
Encompass_Preferred
A secoiridoid glycoside with diverse biological activities; active against E. coli and S. aureus in a disc assay at 2 mg/disc; cytotoxic to human dermal fibroblasts (IC50 = 78.1 µM); prevents hen egg white lysozyme-induced decreases in tail vein blood flow, a marker of allergic inflammation, in hen egg white lysozyme-sensitized mice at 10 mg/kg
Catalog No. 50-298-8958 Supplier Cayman Chemical Supplier No. 3026710 MG
A secoiridoid glycoside with diverse biological activities; active against E. coli and S. aureus in a disc assay at 2 mg/disc; cytotoxic to human dermal fibroblasts (IC50 = 78.1 µM); prevents hen egg white lysozyme-induced decreases in tail vein blood flow, a marker of allergic inflammation, in hen egg white lysozyme-sensitized mice at 10 mg/kg