Learn More
Medchemexpress LLC δ-secretase inhibitor 11 | 842964-18-5 | 99.9% | 220.23 g·mol⁻¹ | C10H12N4O2 | 50MG
SDP
Catalog No. 5000235233
Encompass_Preferred
δ-Secretase inhibitor 11 is a selective, brain-penetrant small-molecule inhibitor of δ-secretase (asparagine endopeptidase) used for neuroscience and neurodegeneration research. It shows reported in vitro potency and is provided as a characterized research-grade compound with supporting analytical data for assay and in vivo studies.
  • Selective δ-secretase inhibition with reported IC50 ≈ 0.7 μM.
  • Brain-penetrant and orally active in preclinical models.
  • High reported purity suitable for research applications.
  • Supplied as a solid with multiple package sizes for experimental flexibility.
  • Comprehensive characterization available, including COA, H-NMR, RP-HPLC, and MS.
Catalog No. 50-002-35233 Supplier Medchemexpress LLC Supplier No. HY12483250MG
δ-Secretase inhibitor 11 is a selective, brain-penetrant small-molecule inhibitor of δ-secretase (asparagine endopeptidase) used for neuroscience and neurodegeneration research. It shows reported in vitro potency and is provided as a characterized research-grade compound with supporting analytical data for assay and in vivo studies.
  • Selective δ-secretase inhibition with reported IC50 ≈ 0.7 μM.
  • Brain-penetrant and orally active in preclinical models.
  • High reported purity suitable for research applications.
  • Supplied as a solid with multiple package sizes for experimental flexibility.
  • Comprehensive characterization available, including COA, H-NMR, RP-HPLC, and MS.