Learn More
Medchemexpress LLC β-secretase inhibitor IV | 797035-11-1 | MFCD09037559 | >98.0% | 578.72 g/mol | C31H38N4O5S | 5 MG
SDP
Catalog No. 5000213461
Encompass_Preferred
β-Secretase Inhibitor IV is a cell-active small-molecule inhibitor of BACE-1 (β-secretase) used in research on amyloid precursor protein processing and Alzheimer's disease pathways. It shows low-nanomolar biochemical potency (IC50 = 15.6 nM at 2 nM BACE-1; 16.3 nM at 100 pM BACE-1). Chemical identifiers: C31H38N4O5S; MW 578.72 g/mol; CAS 797035-11-1.
  • Potent inhibition at low nanomolar concentrations.
  • Demonstrated cell activity in cell-based assays.
  • Suitable for in vitro and cell-based BACE-1 studies.
  • High purity material (>98%) available for research.
  • Small-molecule format compatible with standard assay workflows.
Catalog No. 50-002-13461 Supplier Medchemexpress LLC Supplier No. HY101335MG
β-Secretase Inhibitor IV is a cell-active small-molecule inhibitor of BACE-1 (β-secretase) used in research on amyloid precursor protein processing and Alzheimer's disease pathways. It shows low-nanomolar biochemical potency (IC50 = 15.6 nM at 2 nM BACE-1; 16.3 nM at 100 pM BACE-1). Chemical identifiers: C31H38N4O5S; MW 578.72 g/mol; CAS 797035-11-1.
  • Potent inhibition at low nanomolar concentrations.
  • Demonstrated cell activity in cell-based assays.
  • Suitable for in vitro and cell-based BACE-1 studies.
  • High purity material (>98%) available for research.
  • Small-molecule format compatible with standard assay workflows.