Learn More
Cusabio Technology LLC secreted phosphoprotein 1
Catalog No. 5000049712
Encompass_Preferred
This kit detects OPN in human serum plasma urine and tissue homogenates using a quantitative sandwich ELISA method It offers a broad detection range of 0 78 ng/mL to 50 ng/mL with high sensitivity achieving a lower limit of detection at 0 195 ng/mL The assay requires a sample volume of 50-100 L per test and delivers results within 1-5 hours Optical density measurements are performed at 450 nm to ensure precise quantification of OPN levels Designed for research applications this kit provides reliable performance for analyzing OPN concentrations across diverse biological matrices supporting studies in biomarker discovery disease monitoring and physiological research
Catalog No. 50-000-49712 Supplier Cusabio Technology LLC Supplier No. CSBE08392H
This kit detects OPN in human serum plasma urine and tissue homogenates using a quantitative sandwich ELISA method It offers a broad detection range of 0 78 ng/mL to 50 ng/mL with high sensitivity achieving a lower limit of detection at 0 195 ng/mL The assay requires a sample volume of 50-100 L per test and delivers results within 1-5 hours Optical density measurements are performed at 450 nm to ensure precise quantification of OPN levels Designed for research applications this kit provides reliable performance for analyzing OPN concentrations across diverse biological matrices supporting studies in biomarker discovery disease monitoring and physiological research