Learn More
Medchemexpress LLC HY-P1244A 1mg Medchemexpress, Secretin (33-59), rat (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1244A1MG
Encompass_Preferred
Medchemexpress, HY-P1244A 1mg Secretin (33-59), rat (TFA) CAS: Secretin (33-59), rat (TFA) is a 27-aa peptide, which acts on secretin receptor, and enhances the secretion of bicarbonate, enzymes, and K+ from the pancreas. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0529