Learn More
CPC Scientific H-His-Ser-Asp-Gly-Thr-Phe-Thr-Ser-Glu-Leu-Ser-Arg-Leu-Arg-Glu-Gly-Ala-Arg-Leu-Gln-Arg-Leu-Leu-Gln-Gly-Leu-Val-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific SECT001A
Encompass
Lyophilized solid and gastrointestinal hormone that stimulates the release of a pancreatic, NaHCO3 containing aqueous secretion. This, in turn, neutralizes the hydrochloric acid that has found its way into the small intestine. The gene of secretin is also expressed in brain, heart, kidney, lung, and testis.SEQUENCE: H-His-Ser-Asp-Gly-Thr-Phe-Thr-Ser-Glu-Leu-Ser-Arg-Leu-Arg-Glu-Gly-Ala-Arg-Leu-Gln-Arg-Leu-Leu-Gln-Gly-Leu-Val-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: HSDGTFTSELSRLREGARLQRLLQGLV-NH2MOLECULAR FORMULA: C130H220N44O40MOLECULAR WEIGHT: 3039.45STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [108153-74-8]SYNONYMS: Secretin (human)RESEARCH AREA: GastrointestinalREFERENCES:M. Carlquist et al., IRCS Med. Science, 13, 217 (1985)SKU(s): SECT-001A, SECT-001B
Catalog No. 50-211-0386